Learn More
Kimtech™ A5 Sterile Cleanroom Coveralls
GREENER_CHOICE
Product Code. 11398031
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11398031 Medium Pack of 25
11388031 Small Pack of 25
11308041 Large Pack of 25
11318041 X-Large Pack of 25
11328041 2X-Large Pack of 25
11338041 3X-Large Pack of 25
11348041 4X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11398031 Supplier Kimtech™ Supplier No. 88801
  • ONLINE EXCLUSIVE PRICE
    618.50 €
    296.89 € / Pack of 25
    Save 321.61 €
    51% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Sterile cleanroom coveralls featuring innovative Clean-Don™ technology for simpler aseptic donning. Kimtech™ A5 Sterile Cleanroom Coveralls suitable for EU GMP ISO 5 Grade A sterile cleanrooms, the coveralls are individually vacuum-packed and made from light and breathable fabric.
Assured Compliance:
  • PPE Cat III according to Regulation (EU) 2016/425 (EU)
  • Type 5 and 6 certified
  • Certificate of conformance and sterility by lot available online
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
  • Featuring advanced Clean-Don™ technology for fast, simple and compliant aseptic donning
  • Made from low-lint breathable SMS fabric with a cloth-like feel
  • Contamination protection is ensured through high quality sterility assured material with secure ties
  • Individually vacuum-packed and efficiently double-bagged to save space
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size Medium
Material SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Snap
Seam Type Stitched
Sterility Sterile
Resistance None
Certifications/Compliance ISO 9001
Certifications/Compliance (Europe) EN 13034:2005 - Limited Splash Protection, EN ISO 13982-1:2004 - Particle Protection, CE CAT III, Category III PPE - CE Complex
Latex Free Yes
No. of Pockets None