missing translation for 'onlineSavingsMsg'
Learn More

Kimtech™ A8 Clean Overalls
GREENER_CHOICE

Product Code. p-6976004
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15538881 Small Pack of 25
15548881 Medium Pack of 25
15558881 Large Pack of 25
15568881 X-Large Pack of 25
15578881 2X-Large Pack of 25
15588881 3X-Large Pack of 25
15598881 4X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15538881 Supplier Kimtech™ Supplier No. 47691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. Kimtech™ A8 Clean Overalls are made of a high performance, chemical resistant fabric and come in compressed packaging to save space in the gowning room.

  • High performance chemical resistant fabric
  • Innovative, ultra light fabric: 45 gsm
  • Breathable
  • Highly comfortable
  • Compressed packaging to save space in the gowning room
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Material SMS
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance None
Collar Type Standing
Certifications/Compliance PPE Cat III Type 5/6 (Protection against particles/limited splash protection), EN1149-1:1995 Antistatic garment
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.