missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kinase buffer I (5X)

Product Code. 15444449
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444449 250 mL 250mL
15434449 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444449 Supplier Thermo Scientific™ Supplier No. J61967.AK
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Formulation: 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kinase buffer I (5X)
Physical Form Liquid
Quantity 250 mL
Formulation 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.