missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kinase Tracer 178, 25 μM

Product Code. 10340074
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10340074 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10340074 Supplier Thermo Scientific™ Supplier No. PV5593
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 178 to a kinase is detected by addition of a Eu-labeled anti-tag antibody.

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 178 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.

TRUSTED_SUSTAINABILITY

Specifications

Target Kinases (Multiple)
For Use With (Application) TR-FRET
Assay Kinase Binding Assay
Content And Storage Store at -20°C.
Assay Entry Biochemical binding/kinetic
Label or Dye Alexa Fluor 647
Product Type Kinase Tracer
Quantity 25 μL
Format 384-well plate
Readout Kinetic
Shipping Condition Dry Ice
Target Entry PTK6, DDR2, BMPR1A, RAF1 (cRAF) Y340D Y341D, ACVR1B, WNK2, TEC, DDR1, EPHB4, ERBB4, KIT V654A, TGFBR1, NLK, TGFBR2, EPHB3, BRAF V599E, BRAF, ACVR2B, ACVR2A
Product Line LanthaScreen
For Use With (Equipment) Microplate Reader
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.