missing translation for 'onlineSavingsMsg'
Learn More

Alexa Fluor™ Kinase Tracer 199, 25 μM

Product Code. 10275264
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10275264 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10275264 Supplier Alexa Fluor™ Supplier No. PV5830
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alexa Fluor™ Kinase Tracer 199 is detected by addition of a Eu-labeled anti-tag antibody.

  • Color: Far-Red
  • Excitation/Emission (nm): 340/665
  • Label or Dye: Alexa Fluor™ 647
  • Target Enzyme: Kinases (Multiple)
  • Validated Kinases: EGFR T790M L858R, EPHB4, KIT V654A, MAPK9 (JNK2), RIPK2
  • In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 199 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.
TRUSTED_SUSTAINABILITY

Specifications

Assay Kinase Binding Assay
Assay Entry Biochemical binding/kinetic
Color Far-Red
Content And Storage Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340/665
For Use With (Application) TR-FRET
Label or Dye Alexa Fluor™ 647
Quantity 25 μL
Shipping Condition Dry Ice
Target Entry MAPK8, EGFR, RIPK2, MAPK9, EGFR T790M L858R, MAPK14, MAPK10, EGFR L861Q, EGFR L858R
Type Kinase Tracer
Validated Kinases EGFR T790M L858R, EPHB4, KIT V654A, MAPK9 (JNK2), RIPK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.