Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Alle ansehen Thermo Scientific ProdukteBeschreibung
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Spezifikation
Spezifikation
| Product Type | Tip Comb and Plate |
| Description | KingFisher 96 Deep-Well Tip Comb & Plate, Sterile, Barcoded |
| For Use With (Equipment) | KingFisher Apex |
| Quantity | 50/Cs. |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.