missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex 96 Deep-well Head Magnetic Particle Processor for PrepSEQ Chemistry

Product Code. 17986840 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17986840 Case of 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17986840 Supplier Thermo Scientific™ Supplier No. A57714
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ Pharma KingFisher Flex 96 Deep-well Head Magnetic Particle Processor is intended for pharmaceutical analytics applications and is an excellent option for facilities that wish to automate their PrepSEQ Residual DNA or PrepSEQ 1-2-3 nucleic acid extraction kit protocols.

The Thermo Scientific™ Pharma KingFisher Flex 96 Deep-well Head Magnetic Particle Processor is intended for pharmaceutical analytics applications and is an excellent option for facilities that wish to automate their PrepSEQ Residual DNA or PrepSEQ 1-2-3 nucleic acid extraction kit protocols. The KingFisher Flex platform is a premier automated platform designed to meet your high-throughput needs. It seamlessly incorporates the rapid, reliable, and cost-effective magnetic bead-based extraction of nucleic acids that you expect from PrepSEQ technology with a 96-well automation system. Using magnetic particle separation technology, this system provides excellent reproducibility and quality.

Key product features:

  • Reliable recovery – quantitative and consistent nucleic acid recovery and cross-contamination control
  • Efficient isolation – high-quality and high-throughput isolation of nucleic acids from Mycoplasma, Vesivirus, MMV, and residual DNA
  • Streamlined design – superior performance and cost effectiveness per reaction
  • Modular & versatile application – fully compatible with the PrepSEQ nucleic acid extraction kits

Quantitative & consistent recovery
Quantitative and consistent recovery of DNA from challenging matrices – including samples that have high protein concentration, low pH, or high salt – often presents a technical challenge. The PrepSEQ nucleic acid extraction kits are now adapted for higher-throughput workflows (up to 96 extractions, or 24 samples in triplicate) when used in combination with the KingFisher Flex 96 Deep-well Head Magnetic Particle Processor. The special formulation of magnetic particles provided in the PrepSEQ kits help ensure efficient and specific recovery of DNA from challenging samples.

High quality & throughput isolation
Most automated liquid handling systems move reagents into and out of a single well to perform the different steps of nucleic acid isolation. In contrast, the KingFisher Flex 96 Deep-well Head Magnetic Particle Processor utilizes permanent rods to collect the magnetic beads from solution and releases the beads into a well containing reagents for the next step of isolation. The effectiveness of bead collection and transfer leads to superior washing, elution efficiency, and processing. The KingFisher Flex 96 Deep-well Head Magnetic Particle Processor can process up to 24 samples in triplicate, compared to 3 samples in triplicate using the manual method.

Compatible with PrepSEQ sample preparation kits
The KingFisher Flex 96 Deep-well Head Magnetic Particle Processor has been optimized to work with both the PrepSEQ Residual DNA Sample Preparation Kit and the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit assays. The instrument comes pre-installed with PrepSEQ optimized protocols and does not require any further optimization after installation and validation.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Nucleic Acid Extraction and Cell Separation Instrument
Sample Type Tissue Homogenate
Suitable Downstream Analysis PCR/qPCR, Sequencing, Mass Spec
Plates per Deck 8
Format 96 Deep-well Instrument
Quantity Case of 1

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.