missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats

Product Code. 10357939 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
KingFisher Apex
KingFisher Flex
KingFisher Flex and Presto
Quantity:
100/Cs.
48/Cs.
50/Cs.
80/Cs.
Unit Size:
Pack of 100
Pack of 48
Pack of 50
Pack of 80
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
10357939 KingFisher Flex and Presto 48/Cs. Pack of 48
16532452 KingFisher Apex 50/Cs. Pack of 50
12613126 KingFisher Flex 100/Cs. Pack of 100
12603126 KingFisher Flex 80/Cs. Pack of 80
17160069 KingFisher Apex 100/Cs. Pack of 100
17180069 KingFisher Apex 48/Cs. Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10357939 Supplier Thermo Scientific™ Supplier No. 97002540
Black Friday Starts Here
Use promo code 27984
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.

TRUSTED_SUSTAINABILITY

Specifications

Product Type 96-well Plate
Format 96 Deep-well Plate
Capacity 96 Samples
Content And Storage KingFisher 96 microplate (200μL) (for Flex and Presto)

*Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430).

For Use With (Equipment) KingFisher Flex and Presto
Quantity 48/Cs.
Well Shape Conical
No. of Wells 96
Barcode No
Sterility Non-sterile

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.