missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ PlasmidPro Maxi Processor

Product Code. 30280251 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280251 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280251 Supplier Thermo Scientific™ Supplier No. PPMX1000
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The KingFisher™ PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps.

The KingFisher PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps. Simply pour in the culture and let the instrument handle the rest. In under 75 minutes, this innovative system efficiently executes all essential purification steps, delivering high-quality, endotoxin-free plasmid DNA.

True walkaway automation

The KingFisher PlasmidPro system is fully automated, eliminating the need for pipetting or centrifugation steps. It completes each run in under 75 minutes, allowing you to focus on other tasks.

Exceptional quality

Optimized for maxi-scale plasmid DNA purification from 150 mL of bacterial culture, the system consistently delivers high-quality, advanced, transfection-grade plasmid DNA.

Convenient pre-filled cartridge

The one-piece, pre-filled KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge eliminates setup time and ensures a simple clean-up process, enabling quick turnaround for your next purification.

Effortless installation and setup

The installation process can be completed in minutes on any benchtop, ensuring immediate usability.

Optional AB Assurance Plan

The AB Assurance Plan is a comprehensive repair plan that aims to optimize instrument performance and ensure the availability of critical systems. It includes preventive maintenance, proactive instrument monitoring, and repairs, all aimed at keeping your laboratory operating smoothly.

Additional details

The KingFisher PlasmidPro Maxi Processor is specifically optimized for cultures grown in Luria-Bertani (LB) medium.

At the end of the procedure, the system delivers endotoxin-free plasmid DNA resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Plasmid Prep
Sample Type Bacteria
Suitable Downstream Analysis Advanced Transfection Applications
Volume (Metric) Processing 150 mL
Plates per Deck None
Format Cartridge
Capacity 1 Sample/Run
Dimensions 19.7 x 11.8 x 23.6 in. (50 x 30 x 60 cm)
Display 7 in. Touchscreen
For Use With (Application) Plasmid purification, advanced transfection applications
Ports USB
Purification Target Plasmid DNA
Quantity 1 Ea.
Run Time <75 min.
Samples per Run 1 Sample/Run
Sample Volume 150 mL
Shipping Condition Room temperature
Memory 8 GB
Electrical Requirements 100/240 VAC, 50/60 Hz (nominal), automatic voltage detection
Frequency 50/60 Hz
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.