missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher™ Flex Purification System, KingFisher with 24 Deep-well Head

Product Code. 11819520 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11819520 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11819520 Supplier Thermo Scientific™ Supplier No. 5400640
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processors.

Prepare samples efficiently from a variety of materials with the Thermo Scientific™ KingFisher™ Flex Purification System, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20 minutes.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.

Recommended for:

  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) Processing 200 to 5000 μL
Plates per Deck 24 or 96
Capacity 24 Samples
Description KingFisher 24 DW plate
Collection Efficiency of Particles > 95%
Display START / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / four cursor keys / LCD
For Use With (Application) Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating Temperature From +5°C above ambient temperature to +115°C
Magnetic Rods 24 in one frame; Interchangeable magnet heads
Particle Size (Metric) ca. >1 μm
Quantity Each
Plate Type 24 or 96-well plates: KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 plate (20 to 200 μL), PCR plate (20 to 100 μL)
Depth (English) 23.6 in.
Depth (Metric) 60 cm
Height (English) 15 in.
Height (Metric) 38 cm
Weight (English) 62 lb.
Weight (Metric) 28 kg
Width (English) 26.8 in.
Width (Metric) 68 cm
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.