missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl 4% Boric Acid Solution, without Indicator, Fisher Chemical™

Product Code. 15978866 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15978866 5 L 5 L
15968866 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15978866 Supplier Fisher Chemical Supplier No. K/0205/21
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premade 4% boric acid solution for nitrogen determination by Kjeldahl method.

TRUSTED_SUSTAINABILITY

Specifications

Packaging HDPE Plastic Bottle
Assay Percent Range 3.9 - 4.1%
Quantity 5 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.