missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl 4% Boric Acid Solution, without Indicator, Fisher Chemical™

Product Code. p-8825003 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15968866 1 L 1 L
15978866 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15968866 Supplier Fisher Chemical Supplier No. K/0205/15
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Premade 4% boric acid solution for nitrogen determination by Kjeldahl method.

TRUSTED_SUSTAINABILITY

Specifications

Packaging HDPE Plastic Bottle
Assay Percent Range 3.9 - 4.1%
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.