missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl tablets with selenium (Wieninger)

Product Code. 11984231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11984231 2.5 kg Plastic bottle 2.5kg
10438552 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11984231 Supplier Thermo Scientific Acros Supplier No. 243920025
  • ONLINE EXCLUSIVE PRICE
    466.00 €
    452.00 € / 2.5kg
    Save 14.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-49-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kjeldahl tablets with selenium (Wieninger)
CAS 7782-49-2, 7758-99-8, 7757-82-6
Quantity 2.5 kg
MDL Number MFCD00147338
Packaging Plastic bottle
Product Identifier
  • Kjeldahl tablets with selenium (Wieninger)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.