missing translation for 'onlineSavingsMsg'
Learn More

Kjeldahl Titanium Catalyst Tablets, Technical, Contains 1g Na2SO4, Eq. Of 0.03G Tio2, 0.03G CuSO4, Fisher Chemical

Product Code. 10060250 Shop All Fisher Chemical Products
Click to view available options
:
1000tablets, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10060250

Brand: Fisher Chemical K/0170/81

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Blue
Packaging Plastic powder jar
MDL Number 147340
Quantity 1000 Tablets
Physical Form Solid
compliance-icons
Product Identifier
  • Kjeldahl catalyst tablets Titanium
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.