Learn More
Pacific Handy Cutter Kleen™ Antimicrobial Disposable Film Cutter
Product Code. 17708977
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 50
Product Code. Color Unit Size
17708977 Green Pack of 50
1 options
Product Code. 17708977 Supplier Pacific Handy Cutter Supplier No. DFC364X
  • ONLINE EXCLUSIVE PRICE
    349.32 €
    300.64 € / Pack of 50
    Save 48.68 €
    13% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The KLEEN™ DFC364X is manufactured from a unique, antimicrobial material that inhibits the growth of bacteria on the product. This Disposable Film Cutter features a protective recessed blade, which ensures the highest level of safety.
  • Unlike most utility knives and cutters with exposed blades, the blade in the KLEEN™ DFC364X is set back into the head of the cutter, providing complete protection to the user.
  • For added safety, this convenient tool features a metal bladeless tape splitter which allows users to open boxes without even utilizing a blade, which also helps prevent damaging the contents of boxes.
  • This versatile safety cutter effortlessly slices through stretch/shrink wrap, plastic strapping, bags, box tape, and more.
  • It also features a lanyard hole for tethering to a PHC lanyard for better retention.
Specifications
Product Type Kleen Knife
Disposable Single-use
Sterility Non-sterile
Color Green