Learn More
KleenGuard™ Jackson Safety™ V20 Purity Eyewear
GREENER_CHOICE
Product Code. p-200080452
Click to view available options
Lens Coating:
Anti-fog
UV-treated
Unit Size:
Pack of 12
Product Code. 11735054
Brand: KleenGuard™ 25654
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows you to provide stylish, yet economical eye protection for yourself and your team. KleenGuard™ Jackson Safety™ V20 Purity Eyewear features padded temples for enhanced comfort.
  • Complies with: EN166:2001 1F for safety
  • Complies with: EN166:2001 level F for impact resistance
  • Complies with: EN166:2001 class 1 for optical clarity
  • 99% UVA/UVB protection
  • Comfortable to wear for long periods
  • Economical eye protection for workers and visitors
Specifications
Certifications/Compliance EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) Basic eye protection while keeping costs down, keeping lenses clear and free from condensation during temperature or humidity changes
No. per Pack 12 Pk.
Weight (Metric) 0.264 kg
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless