missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ A50 Breathable Splash and Particle Protection Jacket

Product Code. 12611446
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12611446 Medium Pack of 15
12631446 X-Large Pack of 15
12621446 Large Pack of 15
12641446 2X-Large Pack of 15
12651446 3X-Large Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12611446 Supplier KleenGuard™ Supplier No. 99440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Single-use, breathable jacket with specially designed hood to fit a respirator. KleenGuard™ A50 Breathable Splash and Particle Protection Jacket is designed for comfort, safety and general protection against larger particles, radioactive dust and chemical splashes.

  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
  • Single use and breathable protective clothing designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Silicone-free, ultra-low lint and treated to be antistatic - to EN 1149-5
  • Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals reducing contamination in critical areas
  • Suitable for use in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
  • Storm flap and hood designed for easier taping to a respirator
TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Chemical Resistant Lab Coat
Size Medium
Material Spunbond Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Puncture, Flame, Abrasion, Tears, Particulates, Liquids
Style Full-length
No. of Pockets None
For Use With (Application) Providing high levels of protection against larger particles, radioactive dust and chemical splashes
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.