missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.
Highlights
- Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
- Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers
Applications
- DNA blunting by fill-in 3'-overhangs
- Random-primed DNA labeling
- Labeling by fill-in 5'-overhangs of dsDNA
- DNA sequencing by the Sanger method
- Site-specific mutagenesis of DNA with synthetic oligonucleotides
- Second strand synthesis of cDNA
Specifications
Specifications
| Concentration | 10 U/μL |
| Polymerase | DNA Polymerase I |
| Quantity | 300 units |
| Product Type | Klenow Fragment |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?