missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klercide 70/30 IPA Aerosol Blended with Water for Injection

Product Code. 17885487
Click to view available options
Volume (Metric):
24 x 360 mL
Unit Size:
8.64 L
This item is not returnable. View return policy

Product Code. 17885487

Brand: Ecolab 6600973

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klercide 70/30 IPA blended with the highest grade Water for Injection providing a versatile, high quality product for use in Grade A and B cleanrooms.

  • BPR approved
  • Versatile, high grade disinfection with easy application
  • Triple bagged for easy transfer into Grade A and B cleanrooms
  • 0.2 micron filtered, filled and triple bagged in a Grade C (ISO Class 7) cleanroom
  • Irradiated at no less than 25 kGy to ensure a sterile alcohol
  • Endotoxin tested to an approved pharmacopoeial method
  • Supplied in an aerosol, dispensing a fine mist of alcohol
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available

Specifications

Product Type Disinfectant
Form Aerosol
pH Neutral
Sterility Sterile
Volume (Metric) 24 x 360 mL
compliance-icons compliance-icons
Product Identifier
  • Klercide 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Isopropanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.