Learn More
Ecolab™ Klercide™ Neutral Detergent Unit Dose
Product Code. 15318488
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 50
Product Code. Volume (Metric) unitSize
15318488 100 mL Pack of 50
1 options
Product Code. 15318488 Supplier Ecolab™ Supplier No. 3078810
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The neutral detergent is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
  • The product is a sterile detergent concentrate, to be diluted at a 1:49 ratio, for large surfaces
Specifications
Quantity 50/Pk.
Volume (Metric) 100 mL