missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide Sporicidal Enhanced Peroxide Blended with DI Spray
Description
The Sterishield Delivery System is the only fully validated closed system designed to keep the fluid contents of the bottle sterile at all times. This ensures full compliance with GMP Annex 1 requirements that disinfectant solutions are sterile prior to use.
- BPR approved
- Ready-to-use disinfectant for proactive control of spores, particularly on larger surfaces where safe handling is especially important
- Short contact times against bacteria and fungi even in the presence of background soiling
- Low odor: Safe for operators and cleanroom surfaces
- Effective broad spectrum disinfectant efficacy even when the level of bioburden may be variable, such as during transfer disinfection
- No hazard classification: Free of aldehyde, chorine and peracetic acid
- Broad virucidal efficacy
- Supplied in a 1 L trigger spray incorporating the Sterishield Delivery System, which ensures in-use sterility for up to 3 months
- Double bagged
- Low surface tension properties which ensure an optimum spread of the disinfectant and ideal surface wetting for effective results
- Certificates of Analysis and Sterility are available for every batch
- Full supporting technical documentation is available
Spezifikation
Spezifikation
| Product Type | Cleanroom Biocide |
| Form | Liquid |
| pH | 2.01 to 2.30 |
| Scent | Unscented |
| Sterility | Sterile |
| Appearance | Clear to Light Yellow |
| Composition | 1.5% Hydrogen Peroxide with Deionized Water |
| Container Type | Trigger Bottle |
| Disposable | Single-use |
| Flammable | No |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?