Learn More
Ecolab Klercide WFI Quality Water
Description
- Double bagged for easy transfer into Grade A and B cleanrooms
- Easy to use in a choice of formats
- Cost effective
- Highly effective way of removing any residues after disinfection, simply wipe down with Klercide WFI Quality Water after using the Klercide range of biocides in areas where residues may be an issue.
- Useful for the dilution of sterile concentrates to maintain the required quality of diluted product for high grade cleanrooms
Specifications
Specifications
| Sterility | Sterile |
| Composition | WFI Quality Water |
| Volume (Metric) | 5 L |