missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klerwipe 70/30 IPA Pouch Wipe Blended with Deionized Water

Product Code. 17845507
Change view
Click to view available options
Dimensions (L x W):
300 x 300 mm
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17845507 300 x 300 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17845507 Supplier Ecolab Supplier No. 6600983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Klerwipe 70/30 IPA Tub wipes are sterile, non woven, polyester and cellulose blend wipes impregnated with 70% isopropyl alcohol and blended with deionized water.

  • BPR approved
  • For rapid controlled disinfection of small surfaces and equipment
  • Impregnated with 0.2 μm filtered IPA|DI before being terminally sterilised by gamma irradiated at not less than 25KG
  • Double bagged resealable tub with a low shedding aperture
  • Contains 100 wipes on a perforated roll
  • Wiping can improve the effectiveness of the cleaning and disinfection procedure. Klerwipe are highly absorbent and remain wet throughout use ensuring a consistent application of alcohol to the whole surface area
  • Using impregnated wipes eliminates over-spraying of alcohol, reducing the risk of inhalation, especially in tight working spaces. The alcohol evaporates completely and the wipes do not contain any chemical binders, so no undesirable residues remain
  • Certificates of Analysis are available with every batch

Specifications

Product Type Wipe
Length (Metric) 300 mm
Width (Metric) 300 mm
Material Polyester
Dimensions (L x W) 300 x 300 mm
Sterility Sterile
compliance-icons compliance-icons
Product Identifier
  • 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.