missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klerwipe™ 70/30 IPA Wipes

Product Code. 15316118
Change view
Click to view available options
Dimensions (L x W):
200 x 200 mm
300 x 300 mm
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15316118 200 x 200 mm Pack of 20
15336118 300 x 300 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15316118 Supplier Ecolab™ Supplier No. 3079330

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use in sterile environment. Ecolab™ Klerwipe™ 70/30 IPA Wipes are composed of low particulate or non-woven material.

  • Impregnated with choice of 70% alcohols, either IPA or Denatured Ethanol, blended with deionised water or Water for Injection and choice of biocides
  • Klertear feature makes packs easy to open even when wearing gloves
  • Made from a 22, 44 or 68gsm, binder free, hydroentangled polyester/cellulose blend
  • Impregnated with 70% IPA or Denatured Ethanol
  • Double bagged in resealable pouch or individual laminate sachets and sterilised by gamma irradiation

Specifications

Product Type Impregnated Alcohol Wipes
Dimensions (L x W) 200 x 200 mm
For Use With (Application) Sterile environment
compliance-icons compliance-icons
Product Identifier
  • Klerwipe 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.