missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klerwipe Sporicidal Enhanced Peroxide Blended with DI Mop Wipe
Click to view available options
Dimensions (L x W):
420 x 250 mm
Unit Size:
Pack of 200
Description
- BPR approved
- Ready-to-use disinfectant for proactive control of spores, particularly on larger surfaces where safe handling is especially important
- Sporicidal efficacy
- Short contact times against bacteria and fungi even in the presence of background soiling
- Low odor: Safe for operators and cleanroom surfaces
- Effective broad spectrum disinfectant efficacy even when the level of bioburden may be variable, such as during transfer disinfection
- No hazard classification: Free of aldehyde, chlorine and peracetic acid
- Broad virucidal efficacy
- Mop wipe format provides a practical solution for large surface disinfection in smaller cleanroom areas where bucket systems are difficult to use and create additional maintenance costs
- Easy to open peelable pack
- The product has low surface tension properties which ensure an optimum spread of the disinfectant and ideal surface wetting for effective results
- Certificates of Analysis and Sterility are available for every batch
- Full supporting technical documentation is available
Specifications
Specifications
| Product Type | Cleanroom Biocide |
| pH | 2.01 to 2.30 |
| Scent | Unscented |
| Sterility | Sterile |
| Appearance | Clear to Light Yellow |
| Composition | 1.5% Hydrogen Peroxide with Deionized Water |
| Dimensions (L x W) | 420 x 250 mm |
| Disposable | Single-use |
| Filtered | Filtered |
| Flammable | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction