Learn More
Shield Medicare™ Klerwipe™ Sterile Low Particulate Dry Wipes
Product Code. 12878040
Change view
Click to view available options
Dimensions (L x W):
400 x 400 mm
Unit Size:
Pack of 100
Product Code. Dimensions (L x W) unitSize
12878040 400 x 400 mm Pack of 100
1 options
Product Code. 12878040 Supplier Shield Medicare™ Supplier No. 3035520
Only null left
Add to basket
Add to basket
Low particulate binder-free hydroentangled polyester/cellulose wipes
  • Each pack of wipes, featuring Klertear, is packed in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Specifications
Description Low Particulate Dry Wipe
Dimensions (L x W) 400 x 400 mm
Packaging Quantity 10 Pk. x 10 Wipes