missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Knob Lid

Product Code. 10599151 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outer:
215 mm
270 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10599151 215 mm Each
10401063 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10599151 Supplier BRAND™ Supplier No. 65631
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Knob Lid designed for use with a variety of desiccators.

  • Knob on top of lid
  • Constructed of glass
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 215 mm
Material Borosilicate Glass (3.3)
For Use With (Equipment) Desiccator
Includes Knob
Diameter (Metric) Inner 150 mm
Product Type Desiccator Lid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.