missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Gibco™ KnockOut™ Serum Replacement – Multi-Species (KnockOut SR – Multi-Species) is a more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells. Designed to replace fetal bovine serum (FBS) in existing protocols, this Research Use Only product uses the same KnockOut Serum Replacement formulation that has been trusted by many for almost 20 years. For pre-clinical or clinical applications, please see Cat. No. 10828010.
With KnockOut Serum Replacement – Multi-Species, you can:
Culture mammalian PSCs with confidence
The KnockOut SR – Multi-Species formulation has been cited in more than 2000 publications. It has been shown to support feeder-dependent culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) from mouse, human, and other mammalian species. For mouse and human PSCs, it has also been shown to support reprogramming, embryoid body (EB) formation, in vitro differentiation, cryopreservation, ESC derivation, and many other applications.
Rely on improved performance
KnockOut SR – Multi-Species has a more defined formulation compared to FBS, which contains many undefined and variable components. Unlike most FBS products, each lot is tested for its ability to support PSCs. Because of this, culturing mouse and human PSCs with KnockOut SR – Multi-Species leads to less differentiation and more compact colony morphology compared to FBS.
Enjoy the peace of mind of having stable price and supply
KnockOut SR – Multi-Species is not affected by the limited supply and fluctuating prices often seen with FBS. This makes it easier to plan your budget and buy what you need, especially since it is more affordable than most FBS products.
For Research Use Only. Not for use in diagnostic procedures.
With KnockOut Serum Replacement – Multi-Species, you can:
- Culture mouse, human, and other mammalian feeder-dependent PSCs with confidence
- Rely on the improved performance of a qualified, FBS-free formulation
- Enjoy the peace of mind of having a more stable price and supply compared to FBS
Culture mammalian PSCs with confidence
The KnockOut SR – Multi-Species formulation has been cited in more than 2000 publications. It has been shown to support feeder-dependent culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) from mouse, human, and other mammalian species. For mouse and human PSCs, it has also been shown to support reprogramming, embryoid body (EB) formation, in vitro differentiation, cryopreservation, ESC derivation, and many other applications.
Rely on improved performance
KnockOut SR – Multi-Species has a more defined formulation compared to FBS, which contains many undefined and variable components. Unlike most FBS products, each lot is tested for its ability to support PSCs. Because of this, culturing mouse and human PSCs with KnockOut SR – Multi-Species leads to less differentiation and more compact colony morphology compared to FBS.
Enjoy the peace of mind of having stable price and supply
KnockOut SR – Multi-Species is not affected by the limited supply and fluctuating prices often seen with FBS. This makes it easier to plan your budget and buy what you need, especially since it is more affordable than most FBS products.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Certifications/Compliance | EU-Approved |
| Classification | Serum-Free |
| Content And Storage | Store in freezer (-5 to -20°C) and protect from light. |
| Product Type | Serum Replacement |
| Quantity | 500 mL |
| Sterility | Sterile |
| Manufacturing Quality | cGMP for medical devices, 21 CFR Part 820 and ISO 13485 |
| Lyophilized | Yes |
| Product Line | KnockOut |
| Shipping Condition | Dry Ice |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?