missing translation for 'onlineSavingsMsg'
Learn More

Kojic acid, 100%, MP Biomedicals™

Product Code. 11453352 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453352 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453352 Supplier MP Biomedicals Supplier No. 0215515701
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 501-30-4
Color White
Assay Percent Range 1
Molecular Formula C6H6O4
MDL Number MFCD00006580
Quantity 1 g
Synonym kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
SMILES OCC1=CC(=O)C(O)=CO1
IUPAC Name 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular Weight (g/mol) 142.11
PubChem CID 3840
ChEBI CHEBI:43572
Formula Weight 142.1
Physical Form Crystalline Powder
Chemical Name or Material Kojic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.