missing translation for 'onlineSavingsMsg'
Learn More

Kojic Acid, TRC

Product Code. 30012782
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012782 10 g 10g
30011143 25 g 25g
30003902 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012782 Supplier Toronto Research Chemicals Supplier No. TRCK65508010G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6 H6 O4
Molecular Weight (g/mol) 142.11
Synonym 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 2-(Hydroxymethyl)-5-hydroxy-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyranone, 5-Hydroxy-2-hydroxymethyl-1,4-pyrone, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethylpyran-4-one, Kojic acid, NSC 1942
IUPAC Name 5-hydroxy-2-(hydroxymethyl)pyran-4-one
SMILES OCC1=CC(=O)C(=CO1)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H6O4/c7-2-4-1-5(8)6(9)3-10-4/h1,3,7,9H,2H2
Formula Weight 142.0266
Percent Purity >95%
Chemical Name or Material Kojic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.