Learn More
MEDICOM Kolmi™ Skidguard PP+PE Recycled Overshoes
Product Code. 31781298 Shop All Medicom Products
Change view
Click to view available options
Color:
White
White, Blue
Unit Size:
Pack of 400
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31781298 White, Blue Pack of 400
18886688 White Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31781298 Supplier MEDICOM Supplier No. 10.040R
  • ONLINE EXCLUSIVE PRICE
    58.37 €
    51.93 € / Pack of 400
    Save 6.44 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Recycled KOLMI™ SkidGuard size XL shoe covers provide durable non-slip protection thanks to recycled materials and a high-performance PE sole.
  • The hydrophobic spunbond of the shoe cover is made from 90% recycled-origin material
  • The PE sole incorporates at least 50% recycled-origin material
  • The PE sole ensures good mechanical resistance while providing excellent floor adhesion
  • Their anatomical shapes allow them to adapt to all types of shoes and morphologies, especially when used without shoes, notably in the medical field
Specifications
Size X-Large
Material Polyethylene, Polypropylene
Latex Free Yes
Color White, Blue
Disposable Polyethylene
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Liquid
Product Type Shoe Covers