Learn More
Medicom™ KOLMI Waterproof Overshoes with Non-slip SafeFeet SkidGuard Sole
Product Code. 17100176 Shop All Medicom Products
Change view
Click to view available options
Sole Type:
Anti-slip
Unit Size:
Pack of 400
Product Code. Sole Type Unit Size
17100176 Anti-slip Pack of 400
1 options
Product must be used on intact skin only. The reuse or extended use may lead to infection or cross-contamination.
Product Code. 17100176 Supplier Medicom™ Supplier No. H06711B
  • ONLINE EXCLUSIVE PRICE
    92.66 €
    74.85 € / Pack of 400
    Save 17.81 €
    19% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Product must be used on intact skin only. The reuse or extended use may lead to infection or cross-contamination.
These KOLMI Waterproof Non-slip Overshoes are breathable and lightweight with elasticated ankles.
  • Made of non-woven spunbonded 100% polypropylene
  • Glue sealing
  • Elastic, latex-free ankle
  • Supple and slip resistant
  • Anti-slip 60 μm white sole
Specifications
Size X-Large
Material Polypropylene
Latex Free Yes
Color White, Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-slip
Resistance Water
Product Type Shoe Covers