missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Butyl Rubber Stopper, Plug style
Click to view available options
Stopper Number:
13mm
Unit Size:
Pack of 1000
Description
- Economical alternative for low temperature applications
- Gray butyl rubber
- Lyophillization style
Specifications
Specifications
| For Use With (Application) | Low temperature applications |
| For Use With (Equipment) | Fits GPI aluminum seal finish vials |
| Size | 13 |
| Color | Gray |
| Material | Rubber |
| Product Type | Plug Stopper |
| Stopper Number | 13mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction