Learn More
DWK Life Sciences™ Kimble™ Kontes™ Bubbler Tube, Monier-Williams, 24/40 Joint
Product Code. 17110686 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Unit Size:
Each
Product Code. Standard Taper Joint Unit Size
17110686 24/40 Each
1 options
Product Code. 17110686 Supplier DWK Life Sciences™ Supplier No. 5138020000
  • ONLINE EXCLUSIVE PRICE
    200.55 €
    146.35 € / Pack of 1
    Save 54.20 €
    27% Off
 Restricted Item
Estimated Shipment: 05-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Component Parts of Monier-Williams Sulfites Distillation Apparatus which is used for the determination of free sulfite and a reproducible portion of the bound sulfites in food, such as the carbonyl addition products.
  • Bubbler Tube
  • Standard Taper joint 24/40
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Type Bubbler Tube
Standard Taper Joint 24/40