Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10 with Grids
Product Code. 11982905
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
Unit Size:
Pack of 100
Product Code. Quantity Unit Size
11982905 Each Pack of 100
1 options
Product Code. 11982905 Supplier Kova Plastics by Alltrista Supplier No. 22270141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis. Glasstic Slides are extremely time-efficient and cost-effective.
  • Manufactured from optically clear plastic with same refractive index as glass
  • Each 10-chambered Glasstic Slide features 10 individual fully segregated specimen wells
  • Capillary action reduces pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
  • Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magification
  • Provides results that have high correlation with results obtained with precision hemocytometers: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
Type Glasstic Slide 10:100 Slides with Grids
For Use With (Application) Microscopic examination of Urine and other body fluids
Gridded Yes
Material Plastic with the optical qualities of glass
No. of Wells 10
Quantity Each