Learn More
Klever KS Series Cutter
Product Code. 17739017
Change view
Click to view available options
Blade Material:
Stainless Steel
Unit Size:
Pack of 5
Product Code. Blade Material Unit Size
17739017 Stainless Steel Pack of 5
1 options
Product Code. 17739017 Supplier Klever Supplier No. KS101SSW
  • ONLINE EXCLUSIVE PRICE
    182.12 €
    156.14 € / Pack of 5
    Save 25.98 €
    14% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Klever KS Series Cutter with Wide Opening Stainless Steel Handle and Carriage.
  • Do you want a safe cutting tool that is particularly easy to disassemble because it consists of only two parts (plus blade)? Are you looking for a safety knife that is particularly easy to clean? Then the Klever KS101SS-W safety knife is the one for you.
    • It cuts up to 1.5-wall corrugated cardboard, but also almost any other material.
    • Suitable for auto-clave cleaning
Specifications
Blade Material Stainless Steel
Product Type Cutter
Disposable Reusable
Sterility Non-sterile
Color Silver