missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ KU 55933

Product Code. 18085412
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085412 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18085412 Supplier R&D Systems™ Supplier No. 3544/10
  • ONLINE EXCLUSIVE PRICE
    340.70 €
    307.65 € / 10mg
    Save 33.05 €
    9% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective ATM kinase inhibitor

Potent, selective and competitive ATM kinase inhibitor (Ki = 2.2 nM, IC50 values are 13, 2500, 9300, 16600, > 100000 and > 100000 nM at ATM, DNA-PK, mTOR, PI 3-kinase, PI 4-K and ATR respectively). Decreases viability of MCF-7, A549 and HCT116 cells and decreases p21CIP1 levels in vitro. Acts as a radio- and chemosensitizer for the treatment of cancer.
TRUSTED_SUSTAINABILITY

Specifications

CAS 587871-26-9
Quantity 10 mg
Product Line Tocris
Molecular Formula C21H17NO3S2
Formula Weight 395.49
Purity >98%
Molecular Weight (g/mol) 395.49
Chemical Name or Material KU 55933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.