missing translation for 'onlineSavingsMsg'
Learn More

KU 55933, TRC

Product Code. 30389512
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389512 5 mg -
30389511 10 mg 10mg
30396125 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389512 Supplier Toronto Research Chemicals Supplier No. TRCK6604385MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587871-26-9
Molecular Formula C21H17NO3S2
Molecular Weight (g/mol) 395.49
Synonym KU 55933, ATM Kinase Inhibitor, 2-Thianthren-1-yl-6-(morpholin-4-yl)-4H-pyran-4-one
SMILES O=C1C=C(OC(=C1)c2cccc3Sc4ccccc4Sc23)N5CCOCC5

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H17NO3S2/c23-14-12-16(25-20(13-14)22-8-10-24-11-9-22)15-4-3-7-19-21(15)27-18-6-2-1-5-17(18)26-19/h1-7,12-13H,8-11H2
Chemical Name or Material 2-(4-Morpholinyl)-6-(1-thianthrenyl)-4H-pyran-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.