missing translation for 'onlineSavingsMsg'
Learn More

Klever Kutter Metal Detectable

Product Code. 17718987
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17718987 Blue Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17718987 Supplier Klever Supplier No. KCJ1MD
  • ONLINE EXCLUSIVE PRICE
    301.40 €
    259.81 € / Pack of 50
    Save 41.59 €
    13% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Klever Kutter is our all-rounder! And the forefather of all safety cutters with a concealed blade! Small, handy, versatile, and especially safe!

  • Highest level of security, protects users and goods at the same time.
  • Double blade head, therefore particularly economical.
  • Disposable.
  • Our metal-detectable version, specially designed for the food, pharmaceutical and chemical industries.
  • Ideal for single-wave corrugated cardboard, cardboard, foil, adhesive tape, delivery note bags, strapping tape, paper, string, etc.
  • With practical eyelet for lanyard fastening or hanging.

Specifications

Product Type Cutter Metal Detectable
For Use With (Application) Designed for The Food, Pharmaceutical and Chemical Industries
Disposable Single-use
Sterility Non-sterile
Color Blue
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.