Learn More
Masterflex™ Kynar™ Straight Connector, PVDF
Combine these simple, economical straight connector with flexible tubing for use in your low-pressure applications (generally less than 30 psi). Kynar™ PVDF Straight Connectors slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
Brand:  Masterflex™ 30703-05
Product Code. 15318883
  • ONLINE EXCLUSIVE PRICE
    36.35 €
    34.32 € / Pack of 10
    Save 2.03 €
    5% Off
In Stock
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Designed for quick and easy assembly without tools
  • Excellent chemical resistance
  • Temp range: -62 to 129°C
  • Sterilize by ethylene oxide or autoclave