missing translation for 'onlineSavingsMsg'
Learn More

L(-)-2-Amino-3-phenyl-1-propanol, 98%

Product Code. 10553741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553741 5 g Glass bottle 5g
10419670 1 g Glass bottle 1g
10006283 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10553741 Supplier Thermo Scientific Acros Supplier No. 183870050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3182-95-4 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3182-95-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00004732
InChI Key STVVMTBJNDTZBF-VIFPVBQESA-N
Synonym l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninol
PubChem CID 447213
IUPAC Name (2S)-2-amino-3-phenylpropan-1-ol
SMILES N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 90°C to 94°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2CH(NH2)CH2OH
Quantity 5 g
Specific Rotation -23
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,,soluble in chloroform
Formula Weight 151.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-2-Amino-3-phenyl-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-2-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.