missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-2-Phenylglycine, TRC

Product Code. 17959344
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17959344 Supplier Toronto Research Chemicals Supplier No. TRCP32713025G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2935-35-5
Molecular Formula C8 H9 N O2
Synonym L-(+)-2-Phenylglycine, Benzeneacetic acid, α-amino-, (αS)-, Benzeneacetic acid, α-amino-, (S)-, Glycine, 2-phenyl-, L- (8CI), (αS)-α-Aminobenzeneacetic acid, (+)-Phenylglycine, (2S)-2-Amino-2-phenylacetic acid, (2S)-2-Azaniumyl-2-phenylacetate, (S)-(+)-2-Phenylglycine, (S)-(+)-Phenylglycine, (S)-2-Amino-2-(phenyl)acetic acid, (S)-2-Amino-2-(phenyl)ethanoic acid, (S)-Amino(phenyl)acetic acid, (S)-Phenylglycine, (S)-α-Aminobenzeneacetic acid, L(+)-α-Phenylglycine, L-(+)-Phenylglycine, L-(+)-α-Aminophenylacetic acid, L-2-Phenylglycine, L-α-Aminophenylacetic acid, L-α-Phenylglycine, NSC 206293, S-(+)-α-Phenylglycine
InChI Formula InChI=1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)/t7-/m0/s1
SMILES N[C@H](C(=O)O)c1ccccc1
IUPAC Name (2S)-2-amino-2-phenylacetic acid
Molecular Weight (g/mol) 151.1626
Quantity 25 g
Formula Weight 151.0633
Shipping Condition Room Temperature
Chemical Name or Material L-Phenylglycine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.