missing translation for 'onlineSavingsMsg'
Learn More

L-β-3,4-Dihydroxyphenylalanine, 99%, MP Biomedicals™

Product Code. 11497920 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497920 100 g 100g
11487920 25 g 25g
11407930 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497920 Supplier MP Biomedicals Supplier No. 0210157880
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
PubChem CID 6047
ChEBI CHEBI:15765
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Assay Percent Range 0.99
Quantity 100 g
Formula Weight 197.19
Grade Not Available
Physical Form Crystalline
Chemical Name or Material L-β-3, 4-Dihydroxyphenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.