missing translation for 'onlineSavingsMsg'
Learn More

L-3-Nitrophenylalanine, TRC

Product Code. 30469733
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30469733 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30469733

Brand: Toronto Research Chemicals TRCN502250250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-3-Nitrophenylalanine
CAS 19883-74-0
Molecular Formula C9H10N2O4
Quantity 250 mg
Synonym L-3-(m-nitrophenyl)-alanine, 3-Nitro-L-phenylalanine, NSC 21948
InChI Formula InChI=1S/C9H10N2O4/c10-8(9(12)13)5-6-2-1-3-7(4-6)11(14)15/h1-4,8H,5,10H2,(H,12,13)/t8-/m0/s1
SMILES OC([C@@H](N)CC1=CC(N(=O)=O)=CC=C1)=O
IUPAC Name (S)-2-amino-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol) 210.19
Formula Weight 210.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.