missing translation for 'onlineSavingsMsg'
Learn More

L-4-chlorophenylalanine, 98%, Thermo Scientific™

Product Code. 10511711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511711 100 mg Glass bottle 100mg
10380994 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10511711 Supplier Thermo Scientific Acros Supplier No. 302941000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14173-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00063065,MFCD00079675
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym 4-chloro-l-phenylalanine, l-4-chlorophenylalanine, h-phe 4-cl-oh, s-2-amino-3-4-chlorophenyl propanoic acid, l-p-chlorophenylalanine, 2s-2-amino-3-4-chlorophenyl propanoic acid, fenclonine, s, l-pcpa, unii-snm151oe2c, l-phe 4-cl-oh
PubChem CID 736190
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 235.0°C to 238.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4CH2CH(NH2)CO2H
Quantity 100 mg
Specific Rotation − 27.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
IUPAC Name (2S)-2-amino-3-(4-chlorophenyl)propanoic acid
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-4-Chlorophenylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.