missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Alanine

Product Code. 30017969
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30017969 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30017969 Supplier Melford Supplier No. A20060100.0
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

L-Alanine is a crucial building block for protein synthesis.

L-Alanine is a crucial building block for protein synthesis. It is incorporated into polypeptide chains during translation, contributing to the structure and function of proteins in various biological processes. L-Alanine serves as a precursor for the biosynthesis of other important molecules, including nucleotides and certain coenzymes.

L-Alanine is used in cell culture media in laboratories for the cultivation of cells. Its presence contributes to the overall metabolic pathways within cells and is essential for supporting the growth and viability of cells in artificial environments.

Specifications

Chemical Name or Material L-Alanine
CAS 56-41-7
Molecular Formula C3H7NO2
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.