missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, MedChemExpress

Product Code. 16431348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431348 Supplier Medchem Express Supplier No. HYN0229
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Alanine is a non-essential amino acid, involved in sugar and acid metabolism, increases immunity, and provides energy for muscle tissue, brain, and central nervous system.

L-Alanine is a non-essential amino acid, involved in sugar and acid metabolism, increases immunity, and provides energy for muscle tissue, brain, and central nervous system.

Specifications

Chemical Name or Material L-Alanine
CAS 56-41-7
Color White
Purity Grade Notes Research
Molecular Formula C3H7NO2
Synonym L-2-Aminopropionic acid
Solubility Information H2O : 150 mg/mL (1683.69 mM; Need ultrasonic and warming) ∣DMSO : 1 mg/mL (11.22 mM; ultrasonic and warming and heat to 80°C)
SMILES N[C@@H](C)C(O)=O
Molecular Weight (g/mol) 89.09
Quantity 5 g
Formula Weight 89.09
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.