missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Alanine methyl ester hydrochloride, 99%

Product Code. 10490531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10490531 25 g Glass bottle 25g
10100462 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10490531 Supplier Thermo Scientific Chemicals Supplier No. 308360250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-20-5
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00063663
InChI Key IYUKFAFDFHZKPI-DOIAFFBONA-N
Synonym l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
PubChem CID 2733257
IUPAC Name methyl (2S)-2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](C)N

Specifications

Melting Point 109.0°C to 111.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C3CH(NH2)CO2CH3·HCl
Quantity 25 g
Specific Rotation + 7.00
Formula Weight 139.58
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Alanine methyl ester hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.