missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, Multi-Compendial, J.T. Baker™

Product Code. 15598884 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15598884 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598884 Supplier Avantor J.T.Baker Supplier No. 625306

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56-41-7
Molecular Formula C3H7NO2
Packaging Wide Mouth Amber Glass Bottle
MDL Number MFCD00064410
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES C[C@H](N)C(O)=O
IUPAC Name (2S)-2-aminopropanoic acid
Molecular Weight (g/mol) 89.09
Quantity 1 kg
PubChem CID 5950
ChEBI CHEBI:16977
Formula Weight 89.09
Specific Gravity 1L = 1.40kg
Grade Multi-Compendial
Chemical Name or Material L-Alanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.