missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, TRC

Product Code. 30134594
Change view
Click to view available options
Quantity:
1 g
100 g
250 g
Unit Size:
100g
1g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134594 250 g 250g
30133790 100 g 100g
17968144 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134594 Supplier Toronto Research Chemicals Supplier No. TRCA481500250G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
Synonym Alanine, L- (7CI,8CI), (2S)-2-Aminopropanoic acid, (S)-(+)-Alanine, (S)-2-Aminopropanoic acid, (S)-Alanine, 2-Aminopropionic acid, Alanine, L-(+)-Alanine, L-2-Aminopropanoic acid, L-2-Aminopropionic acid, L-α-Alanine, L-α-Aminopropionic acid, Lactamine, NSC 206315, Propanoic acid, 2-amino-, (S)-, α-Alanine, α-Aminopropionic acid
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1
Quantity 250 g
Formula Weight 89.0477
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Alanine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.